N,N-Diphenylcarbamoyl chloride 98%

Danger

Synonyms: Diphenylcarbamyl chlorideDiphenylcarbamoyl chloride

AC16714-5000 AC16714-1000 AC16714-0250
200018-868EA 46.9 USD
200018-868 103526-196 200012-162
N,N-Diphenylcarbamoyl chloride 98%
N,N-Diphenylcarbamoyl chloride
Formula: C₁₃H₁₀ClNO
MW: 231.68 g/mol
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00000633
CAS Number: 83-01-2
EINECS: 201-450-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR