1-Benzyl-4-hydroxypiperidine 97%
|
Warning
|
Synonyms:
1-Benzylpiperidin-4-ol
AC16683-0250
AC16683-0050
103510-880EA
27.1
USD
103510-880
200027-792
1-Benzyl-4-hydroxypiperidine 97%
1-Benzyl-4-hydroxypiperidine
Formula:
C₁₂H₁₇NO MW: 191.27 g/mol Boiling Pt: 127…128 °C (2 mmHg) Melting Pt: 60…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006503 CAS Number: 4727-72-4 EINECS: 225-226-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...