4-Methoxyphenylhydrazinium chloride 98%

Warning

Synonyms: 4-Methoxyphenylhydrazine hydrochloride

AC16649-0100 AC16649-0500
200017-624EA 109 USD
200017-624 200023-892
4-Methoxyphenylhydrazinium chloride 98%
4-Methoxyphenylhydrazinium chloride
Formula: C₇H₁₁ClN₂O
MW: 174.63 g/mol
Melting Pt: 153…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00012945
CAS Number: 19501-58-7
EINECS: 243-115-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR