Methyl carbazate 97%
|
Warning
|
Synonyms:
Methyl hydrazinocarboxylate
, Methyl hydrazinecarboxylate
AC16614-5000
AC16614-0025
AC16614-1000
200031-902EA
54.2
USD
200031-902
200043-910
103531-582
Methyl carbazate 97%
Methyl carbazate
Formula:
C₂H₆N₂O₂ MW: 90.08 g/mol Boiling Pt: 108 °C (12 mmHg) Melting Pt: 68…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007594 CAS Number: 6294-89-9 EINECS: 228-560-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...