(±)-2,3-Epoxypropyl methacrylate 97% stabilized

Danger

Synonyms: Oxiran-2-ylmethyl methacrylateGlycidyl methacrylate

AC16589-0025 AC16589-1000 AC16589-5000 AC16589-0050
103526-166EA 104 USD
103526-166 200057-700 103526-164 103526-162
(±)-2,3-Epoxypropyl methacrylate 97% stabilized
(±)-2,3-Epoxypropyl methacrylate
Formula: C₇H₁₀O₃
MW: 142.15 g/mol
Boiling Pt: 189 °C (760 mmHg)
Melting Pt: –41 °C
Density: 1.07 g/cm³
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005137
CAS Number: 106-91-2
UN: 2922
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR