Isopentyl nitrite (mixed isomers ) 97% stabilized

Danger

Synonyms: Isoamyl nitrite

AC16570-1000 AC16570-5000 AC16570-0025
103526-156EA 615 USD
103526-156 200057-896 200057-898
Isopentyl nitrite (mixed isomers ) 97% stabilized
Isopentyl nitrite (mixed isomers )
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 96…99 °C (760 mmHg)
Density: 0.87 g/cm³
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002057
CAS Number: 110-46-3
UN: 1113
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR