1,4'-Bipiperidine 95% tech.

Warning

Synonyms: 4-Piperidinopiperidine

AC16545-0500
200010-626EA 409.4 USD
200010-626
1,4'-Bipiperidine 95% tech.
1,4'-Bipiperidine
Formula: C₁₀H₂₀N₂
MW: 168.28 g/mol
Melting Pt: 62…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00006475
CAS Number: 4897-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR