4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) 98%

Danger

Synonyms: Hexahydro-4-methylphthalic anhydride

AC16539-0025 AC16539-5000
200037-596EA 96.7 USD
200037-596 200037-594
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) 98%
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres)
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 120 °C (1mbar)
Melting Pt: –29 °C
Density: 1.16 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005927
CAS Number: 19438-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR