L(+)-Glutamic acid hydrochloride 99%

Danger

Synonyms: (S)-2-Aminopentanedioic acid hydrochloride

AC16523-5000 AC16523-1000
200019-460EA 22.8 USD
200019-460 200043-732
L(+)-Glutamic acid hydrochloride 99%
L(+)-Glutamic acid hydrochloride
Formula: C₅H₁₀ClNO₄
MW: 183.59 g/mol
Melting Pt: 191…193 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012619
CAS Number: 138-15-8
EINECS: 205-315-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR