5-Nitro-2-furaldehyde diacetate 98%

Synonyms: 5-Nitrofurfural Diacetyl Acetal2-(Diacetoxymethyl)-5-nitrofuran5-Nitrofurfurylidene Diacetate(5-Nitrofuran-2-yl)methylene diacetate

AC16513-0025 AC16513-5000 AC16513-1000
103465-630EA 439 USD
103465-630 103465-632 200030-132
5-Nitro-2-furaldehyde diacetate 98%
5-Nitro-2-furaldehyde diacetate
Formula: C₉H₉NO₇
MW: 243.17 g/mol
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00003244
CAS Number: 92-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR