Adenosine 5'-diphosphate disodium salt (ADP disodium salt) hydrate 98%

AC16467-2500 AC16467-0050 AC16467-0010
200010-984EA 86.4 USD
200010-984 103516-532 200030-298
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) hydrate 98%
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) dihydrate
Formula: C₁₀H₁₃N₅Na₂O₁₀P₂·xH₂O
MW: 471.167 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00150927
CAS Number: 16178-48-6
EINECS: 240-314-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR