trans-Benzylideneacetone 99%

Warning

Synonyms: trans-4-Phenyl-3-buten-2-one(E)-4-Phenylbut-3-en-2-one

AC16452-0500 AC16452-1000 AC16452-5000
103510-828EA 42.2 USD
103510-828 103510-830 200027-042
trans-Benzylideneacetone 99%
trans-Benzylideneacetone
Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 260…262 °C (760 mmHg)
Melting Pt: 38.5…41 °C
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00008779
EINECS: 217-587-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR