Diethyl cyclopropane-1,1-dicarboxylate 97%

Synonyms: Diethyl 1,1-cyclopropanedicarboxylateDiethyl 1,1-cyclopropane dicarboxyliate

AC16445-0250
200018-766EA 150.3 USD
200018-766
Diethyl cyclopropane-1,1-dicarboxylate 97%
Diethyl cyclopropane-1,1-dicarboxylate
Formula: C₉H₁₄O₄
Boiling Pt: 94…96 °C (10 mmHg)
Density: 1.05 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00001283
CAS Number: 1559-02-0
EINECS: 216-321-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR