Bathophenanthrolinedisulfonic acid disodium salt hydrate 98%

Synonyms: 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate4,7-Diphenyl-1,10-phenanthrolinedisulphonic acid disodium salt hydrate

AC16405-0050 AC16405-0010
200001-498EA 128 USD
200001-498 103516-530
Bathophenanthrolinedisulfonic acid disodium salt hydrate 98%
Bathophenanthrolinedisulphonic acid disodium salt hydrate
Formula: C₂₄H₁₄N₂Na₂O₆S₂·H₂O
MW: 536.48 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149310
CAS Number: 52746-49-3
EINECS: 258-152-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR