Adenosine 99+%

Synonyms: Adenine riboside(2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diolAdenine-9-β-D-ribofuranoside9-β-D-Ribofuranosyladenine

AC16404-0250 AC16404-0050
200001-496EA 84.7 USD
200001-496 200005-666
Adenosine 99+%
Adenosine
Formula: C₁₀H₁₃N₅O₄
MW: 267.24 g/mol
Melting Pt: 233…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00005752
CAS Number: 58-61-7
EINECS: 200-389-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR