5-Acetoxymethylfurfural 97%

Warning

Synonyms: 5-Formylfurfuryl Acetate5-Acetoxymethyl-2-furaldehyde(5-Formylfuran-2-yl)methyl acetateAcetic Acid 5-Formylfurfuryl Ester

AC16340-0020 AC16340-0100
200042-204EA 40.9 USD
200042-204 200030-030
5-Acetoxymethylfurfural 97%
5-Acetoxymethylfurfural

Appearance: White to beige to brown Powder or crystals and/or chunks

Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 52…56 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00003233
CAS Number: 10551-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR