ɑ-Cyano-3-hydroxycinnamic acid 98%

Warning

Synonyms: 2-Cyano-3-(3-hydroxyphenyl)acrylic Acid

AC16309-0050 AC16309-0250 AC16309-1000
200018-096EA 57.9 USD
200018-096 200048-476 200036-502
ɑ-Cyano-3-hydroxycinnamic acid 98%
alpha-Cyano-3-hydroxycinnamic acid

Appearance: Yellow to yellow-green Crystalline powder

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 230…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00004203
CAS Number: 54673-07-3
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR