Iminostilbene 97%

Synonyms: 5H-Dibenzo[b,f]azepine

AC16254-1000 AC16254-0100
200025-560EA 75.5 USD
200025-560 200037-640
Iminostilbene 97%
Iminostilbene
Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Melting Pt: 197…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00005071
CAS Number: 256-96-2
EINECS: 205-970-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR