4,6-Dichloro-2-(methylthio)pyrimidine 98%

Danger

AC16303-0050 AC16303-0250
200010-134EA 66.8 USD
200010-134 103531-522
4,6-Dichloro-2-(methylthio)pyrimidine 98%
4,6-Dichloro-2-(methylthio)pyrimidine
Formula: C₅H₄Cl₂N₂S
MW: 195.07 g/mol
Boiling Pt: 135…136 °C (14 mmHg)
Melting Pt: 38…42 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006086
CAS Number: 6299-25-8
EINECS: 228-577-6
UN: 3263
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR