Diethyl dimethylmalonate 97%

Synonyms: Diethyl 2,2-dimethylmalonate

AC16266-0500 AC16266-0100
200030-960EA 76.2 USD
200030-960 103510-792
Diethyl dimethylmalonate 97%
Diethyl dimethylmalonate
Formula: C₉H₁₆O₄
MW: 188.2200 g/mol
Boiling Pt: 192 °C (760 mmHg)
Density: 0.99 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00009128
EINECS: 216-582-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR