Diglycolic acid 98%

Warning

Synonyms: 2,2'-Oxydiacetic acid

AC16226-5000 AC16226-0025 AC16226-1000
200031-000EA 229 USD
200031-000 200025-036 103526-086
Diglycolic acid 98%
Diglycolic acid
Formula: C₄H₆O₅
MW: 134.09 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00004309
CAS Number: 110-99-6
EINECS: 203-823-5
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR