1,3-Diaminoguanidine hydrochloride 98%

Warning

Synonyms: N,N'-Diaminoguanidine monohydrochloride

AC16240-0050 AC16240-0250
200019-916EA 100 USD
200019-916 200050-028
1,3-Diaminoguanidine hydrochloride 98%
1,3-Diaminoguanidine hydrochloride

Appearance: White to light beige Granular powder

Formula: CH₇N₅
Melting Pt: 178…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00012948
CAS Number: 36062-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR