Bathocuproin 98%

Warning

Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

AC16134-0010
200009-098EA 80.3 USD
200009-098
Bathocuproin 98%
Bathocuproin
Formula: C₂₆H₂₀N₂
MW: 360.46 g/mol
Melting Pt: 279…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00004972
CAS Number: 4733-39-5
EINECS: 225-240-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR