Isobutyryl chloride 98%

Danger

AC16066-0100 AC16066-2500 AC16066-0010
200006-728EA 64.9 USD
200006-728 200006-726 200012-846
Isobutyryl chloride 98%
Isobutyryl chloride
Formula: C₄H₇ClO
MW: 106.55 g/mol
Melting Pt: –90 °C
Density: 1.017 g/cm³
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000717
CAS Number: 79-30-1
EINECS: 201-194-1
UN: 2395
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR