Hydroxyacetone, Technical Grade

Warning

AC16034-1000 AC16034-5000
102774-128EA 36 USD
102774-128 102775-726
Hydroxyacetone, Technical Grade
Hydroxyacetone
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –17 °C
Density: 1.08 g/cm³
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004669
CAS Number: 116-09-6
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR