2,4,6-Trimethylbenzyl chloride 98%

Danger

Synonyms: α2-Chloroisodurenealpha-2-Chloroisodureneα-2-Chloroisodureneα-Chloro-1,2,4,6-tetramethylbenzene1-(Chloromethyl)-2,4,6-trimethylbenzene

AC15978-0050 AC15978-0250
200030-354EA 92.9 USD
200030-354 200018-072
2,4,6-Trimethylbenzyl chloride 98%
2,4,6-Trimethylbenzyl chloride
Formula: C₁₀H₁₃Cl
MW: 168.67 g/mol
Boiling Pt: 114…115 °C (10 mmHg)
Melting Pt: 34…38 °C
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000901
CAS Number: 1585-16-6
EINECS: 216-440-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR