2,4-Thiazolidinedione 99%

Warning

Synonyms: 1,3-Thiazolidine-2,4-dione

AC15949-1000 AC15949-0250 AC15949-5000
200009-020EA 62.7 USD
200009-020 200004-892 103509-018
2,4-Thiazolidinedione 99%
2,4-Thiazolidinedione
Formula: C₃H₃NO₂S
MW: 117.13 g/mol
Boiling Pt: 178…179 °C (19 mmHg)
Melting Pt: 123…126 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00005478
CAS Number: 2295-31-0
EINECS: 218-941-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR