N-Boc-glycine (boc-gly-OH) 99+%

Danger

Synonyms: Boc-Gly-OH

AC15880-0100
200001-408EA 51.9 USD
200001-408
N-Boc-glycine (boc-gly-OH) 99+%
N-Boc-glycine (Boc-Gly-OH)
Formula: C₇H₁₃NO₄
MW: 175.2 g/mol
Melting Pt: 87…90 °C
Storage Temperature: Refrigerator
EINECS: 224-864-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR