2-Hydroxyhippuric acid 95%

Warning

Synonyms: N-(2-Hydroxybenzoyl)glycine

AC15862-0100 AC15862-0025
200016-604EA 97 USD
200016-604 200022-720
2-Hydroxyhippuric acid 95%
2-Hydroxyhippuric acid
Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 164…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002695
EINECS: 207-661-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR