1-Amino-4-naphthol hydrochloride 90%, Technical Grade

Warning

Synonyms: 4-Amino-1-naphthol hydrochloride

AC15851-0050 AC15851-0250
200010-166EA 220 USD
200010-166 103508-986
1-Amino-4-naphthol hydrochloride 90%, Technical Grade
1-Amino-4-naphthol hydrochloride
Formula: C₁₀H₁₀ClNO
MW: 195.65
Melting Pt: 273 °C (dec.)
Storage Temperature: Ambient
EINECS: 227-727-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR