Cyclohexyl isocyanide 99%

Warning

AC15840-0250 AC15840-0010 AC15840-0050
103525-962EA 116 USD
103525-962 103525-964 200006-160
Cyclohexyl isocyanide 99%
Cyclohexyl isocyanide
Formula: C₇H₁₁N
Boiling Pt: 173…176 °C (760 mmHg)
Density: 0.89 g/cm³
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003839
CAS Number: 931-53-3
EINECS: 213-238-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR