n-Butyl isocyanide 98+%

Danger

AC15838-0050 AC15838-0010
200013-588EA 210 USD
200013-588 200020-018
n-Butyl isocyanide 98+%
n-Butyl isocyanide
Formula: C₅H₉N
Density: 0.79 g/cm³
Storage Temperature: Refrigerator
EINECS: 220-458-7
UN: 1935
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR