2-Amino-5-methyl-1,3,4-thiadiazole 97%

AC15837-0050
200009-148EA 46.8 USD
200009-148
2-Amino-5-methyl-1,3,4-thiadiazole 97%
2-Amino-5-methyl-1,3,4-thiadiazole
Formula: C₃H₅N₃S
Melting Pt: 224…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00003110
CAS Number: 108-33-8
EINECS: 203-573-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR