Tryptamine hydrochloride 98%
|
Warning
|
AC15798-0050
AC15798-1000
AC15798-0100
200033-130EA
70.7
USD
200033-130
200051-104
200051-106
Tryptamine hydrochloride 98%
Tryptamine hydrochloride
Formula:
C₁₀H₁₃ClN₂ Melting Pt: 253…255 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012682 CAS Number: 343-94-2 EINECS: 206-446-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...