4-Methylphenethylamine 97%

Danger

Synonyms: 2-(p-Tolyl)ethylamine

AC15788-0010 AC15788-0050
103531-468EA 150 USD
103531-468 200021-988
4-Methylphenethylamine 97%
4-Methylphenethylamine
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 214 °C (760 mmHg)
Density: 0.93 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00008195
CAS Number: 3261-62-9
EINECS: 221-865-2
UN: 2922
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR