2-Piperidineethanol 95%

Danger

Synonyms: 2-(2-Hydroxyethyl)piperidine

AC15761-0025 AC15761-0050 AC15761-1000 AC15761-5000
103531-462EA 1685.6 USD
103531-462 200009-584 103531-464 200009-586
2-Piperidineethanol 95%
2-Piperidineethanol

Appearance: White to light yellow Low melting crystalline solid

Formula: C₇H₁₅NO
MW: 129.20 g/mol
Boiling Pt: 234 °C (760 mmHg)
Melting Pt: 38…43 °C
Density: 1.01 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00005989
CAS Number: 1484-84-0
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR