(±)-Tetrahydrofurfurylamine 97%

Warning

Synonyms: 2-Tetrahydrofurfurylamine

AC15782-5000 AC15782-1000 AC15782-0050
200020-868EA 19.4 USD
200020-868 200038-896 200020-866
(±)-Tetrahydrofurfurylamine 97%
(±)-Tetrahydrofurfurylamine
Formula: C₅H₁₁NO
Boiling Pt: 153…154 °C (744 mmHg)
Density: 0.98 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00005373
CAS Number: 4795-29-3
EINECS: 225-351-9
UN: 2943
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR