4-Chloro-DL-phenylalanine 98%

Synonyms: H-DL-Phe(4-Cl)-OHp-Chloro-DL-Phenylalanine

AC15728-0050 AC15728-0250
200037-276EA 265 USD
200037-276 200031-148
4-Chloro-DL-phenylalanine 98%
4-Chloro-DL-phenylalanine
Formula: C₉H₁₀ClNO₂
MW: 199.6 g/mol
Melting Pt: >240 °C
Storage Temperature: Ambient
EINECS: 231-051-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR