2-Furohydrazide 98%

Warning

Synonyms: 2-Furoic hydrazide2-Furoic acid Hydrazide2-Furoic acid hydrazide2-Furoylhydrazine2-Furancarboxylic acid Hydrazide2-Furoic Hydrazide2-Furancarboxylic acid hydrazide

AC15716-0250 AC15716-0050
200034-818EA 33.4 USD
200034-818 200046-892
2-Furohydrazide 98%
2-Furohydrazide
Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00003235
CAS Number: 3326-71-4
EINECS: 222-046-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR