Methyl-2-furoate 98%

Danger

Synonyms: Methyl furan-2-carboxylateMethyl 2-Furancarboxylate

AC15687-1000 AC15687-0250 AC15687-5000
200013-222EA 73.2 USD
200013-222 200019-668 103531-454
Methyl-2-furoate 98%
Methyl 2-furoate
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 181 °C (760 mmHg)
Density: 1.176 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00003236
CAS Number: 611-13-2
EINECS: 210-254-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR