4-(2-Keto-1-benzimidazolinyl)piperidine 97%

Warning

Synonyms: 1,3-Dihydro-1-(piperidin-4-yl)-2H-benzimidazol-2-one

AC15670-0050 AC15670-0010
200017-316EA 215 USD
200017-316 200017-314
4-(2-Keto-1-benzimidazolinyl)piperidine 97%
4-(2-Keto-1-benzimidazolinyl)piperidine
Formula: C₁₂H₁₅N₃O
MW: 217.272 g/mol
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00005714
CAS Number: 20662-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR