4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 97%

Warning

Synonyms: (Chloro(4-methoxyphenyl)methylene)dibenzeneMmt-Cl

AC15656-0250
200005-376EA 95 USD
200005-376
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 97%
4-Methoxytrityl chloride (p-Anisylchlorodiphenylmethane)
Formula: C₂₀H₁₇ClO
MW: 308.8 g/mol
Melting Pt: 121…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000814
CAS Number: 14470-28-1
EINECS: 238-463-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR