1-Methyl-4-piperidone 98%

Danger

Synonyms: N-Methyl-4-piperidone

AC15692-1000 AC15692-5000
200040-316EA 246 USD
200040-316 200008-720
1-Methyl-4-piperidone 98%
1-Methyl-4-piperidone
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 55…58 °C (10 mmHg)
Density: 0.98 g/cm³
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006191
CAS Number: 1445-73-4
EINECS: 215-895-5
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR