Homophthalic anhydride 98%

Warning

AC15630-0050 AC15630-0250
200019-328EA 162.62 USD
200019-328 200019-330
Homophthalic anhydride 98%
Homophthalic anhydride
Formula: C₉H₆O₃
Melting Pt: 140…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006894
CAS Number: 703-59-3
EINECS: 211-873-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR