Fumaric acid monoethyl ester 95%

Warning

Synonyms: (E)-4-Ethoxy-4-oxobut-2-enoic acidEthyl hydrogen fumarate

AC15620-0250 AC15620-1000
103510-650EA 67.7 USD
103510-650 200025-430
Fumaric acid monoethyl ester 95%
Fumaric acid monoethyl ester
Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 147 °C (16 mmHg)
Melting Pt: 64…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00002699
CAS Number: 2459-05-4
EINECS: 219-544-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR