8-Bromoadenosine 98%

Synonyms: 8-Bromoadenine-9-β-D-ribofuranoside6-Amino-8-bromopurine riboside

AC15578-2500 AC15578-0010 AC15578-0050
200048-350EA 117 USD
200048-350 103510-634 200005-590
8-Bromoadenosine 98%
8-Bromoadenosine
Formula: C₁₀H₁₂BrN₅O₄
MW: 346.15 g/mol
Melting Pt: 210…212 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005733
CAS Number: 2946-39-6
EINECS: 220-959-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR