Dibutyl phosphonate P 14.5-16%

Warning

Synonyms: Dibutyl phosphiteDi-n-butylphosphite

AC15506-1000 AC15506-5000
200037-042EA 106 USD
200037-042 200037-040
Dibutyl phosphonate P 14.5-16%
Dibutyl phosphonate
Formula: C₈H₁₉O₃P
MW: 194.21 g/mol
Boiling Pt: 118…119 °C (11 mmHg)
Melting Pt: –57 °C
Density: 0.995 g/cm³
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00066633
CAS Number: 1809-19-4
EINECS: 217-316-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR