2'-Deoxyadenosine monohydrate 99%

Warning

Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate

AC15499-0250 AC15499-0050 AC15499-0010
200022-612EA 137 USD
200022-612 200028-694 103510-622
2'-Deoxyadenosine monohydrate 99%
2'-Deoxyadenosine monohydrate
Formula: C₁₀H₁₃N₅O₃·1H₂O
MW: 269.26 g/mol (251.24 anhy)
Melting Pt: 185…187 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149364
CAS Number: 16373-93-6
EINECS: 213-488-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR