Cyclopentylacetic acid 97%

Warning

Synonyms: 2-Cyclopentylacetic acid

AC15487-0100 AC15487-0250
200011-852EA 191 USD
200011-852 200024-874
Cyclopentylacetic acid 97%
Cyclopentylacetic acid
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 133…134 °C (23 mmHg)
Density: 1.02 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00001387
CAS Number: 1123-00-8
EINECS: 214-368-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR