2-Coumaranone 97%

Warning

Synonyms: Benzo[b]furan-2(3H)-oneBenzofuran-2(3H)-one

AC15446-0050 AC15446-0010 AC15446-0100
200022-602EA 36.6 USD
200022-602 200034-738 200046-808
2-Coumaranone 97%
2-Coumaranone
Formula: C₈H₆O₂
MW: 134.13 g/mol
Melting Pt: 47…52 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
EINECS: 209-052-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR